Learn More
SCHOTT AG Lighting and Imaging™ Color Filter for Spotlight
For use with EasyLED single or double Spotlight Plus. Available in Red, Blue, Green or Yellow.
Brand:  SCHOTT AG Lighting and Imaging™ 600680
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15743052
  • € 38.63 / 1 unit
Estimated Shipment
03-12-2020
Add to basket
Description
Description
  • Color filters for Spotlight and EasyLED
  • Use for fixing at spot light with thread
  • Available colors: red, blue, green, yellow.
Specifications
Specifications
EasyLED Single, Double Spotlight Plus
Yellow
Stereo Microscopy Illumination
Color Filter