Learn More
SCHOTT AG Lighting and Imaging™ Color Filter for Spotlight
Color Filter for use with EasyLED single or double Spotlight Plus. Available in Red, Blue, Green, Yellow or Halogen and a polarization filter.
Brand: SCHOTT AG Lighting and Imaging™ 600670
Additional Details : Weight : 5.00000kg
Description
- Color filters for EasyLED single or double Spotlight Plus
- Use for fixing at spot light with thread.
- Available colors: red, blue, green, yellow and halogen.
- Polarization filter available.
Specifications
Color Filter | |
Convert neutral white LED light to green light | |
EasyLED Spotlight Plus (Single or Double) |
Green | |
Stereo Microscopy Illumination |