Description
- Color filters for EasyLED single or double Spotlight Plus
- Use for fixing at spot light with thread.
- Available colors: red, blue, green, yellow and halogen.
- Polarization filter available.
Specifications
Specifications
| Type | Color Filter |
| Color | Red |
| Description | Convert neutral white LED light to red light |
| For Use With (Application) | Stereo Microscopy Illumination |
| For Use With (Equipment) | EasyLED Spotlight Plus (Single or Double) |