missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COM1/p8 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23626AF350
This item is not returnable.
View return policy
Description
COM1/p8 Polyclonal antibody specifically detects COM1/p8 in Human samples. It is validated for Western Blot
Specifications
| COM1/p8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| Candidate of metastasis 1, COM1P8, nuclear protein 1, nuclear protein, transcriptional regulator, 1, p8, Protein p8 | |
| Reacts with the N terminus of the human p8 protein. | |
| 0.1 mL | |
| Apoptosis | |
| 26471 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction