Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
- Supplied as standard
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Mini SDS PAGE, Native PAGE, Gradient, Second Dimension and Nucleic Acid Separations | |
Comb | |
1 Each | |
25 μL |
OmniPAGE Mini Vertical Gel Electrophoresis Unit | |
16 | |
1 mm |