Learn More
Fisherbrand™ Combs for omniPAGE Mini Vertical Gel Electrophoresis Unit, 0.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
€ 18.85
Specifications
Type Comb
Thickness (Metric) 0.5 mm
View More Specs
Products 8
Product Code Brand No. of Wells Volume (Metric) Well Price Quantity & Availability  
Product Code Brand No. of Wells Volume (Metric) Well Price Quantity & Availability  
11857583
View Documents Promotion Details
Fisherbrand™
10 20 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11837593
View Documents Promotion Details
Fisherbrand™
12 16 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11887593
View Documents Promotion Details
Fisherbrand™
16 12.5 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11837603
View Documents Promotion Details
Fisherbrand™
20 10 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11887603
View Documents Promotion Details
Fisherbrand™
5 50 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11837613
View Documents Promotion Details
Fisherbrand™
8 30 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11887613
View Documents Promotion Details
Fisherbrand™
9 25 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
11837583
View Documents Promotion Details
Fisherbrand™
1 Prep, 1 Marker 330 μL
€ 18.85
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Accessory for use with omniPAGE mini vertical gel electrophoresis unit
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Comb
0.5 mm