Learn More
Fisherbrand™ Combs for Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis, 2mm Thick
For use with Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis units.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11877813
Quantity Price
1 € 26.54 / Each
10 € 25.19 / Each
Estimated Shipment: 20-08-2021
Add to basket
Description
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • 18-sample and 36-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Blue
Comb
1 Each
3000 μL
Verti-Gel Maxi, 2-Gel Standard Vertical Gel Electrophoresis Units
1 Prep, 1 Marker
2 mm