Learn More
Lovibond™ Combi pack aluminum #1 / #2
Product Code. 15371905
Click to view available options
Quantity:
100 Tests
Unit Size:
Pack of 100
Product Code. 15371905
Brand: Lovibond™ 517601BT
Tablet reagent test set
Combi pack aluminum #1 / #2 100 tests measuring range 0.01 - 0.30 mg/L ECR method for comparators and photometers. 
Specifications
Quantity 100 Tests
Range Aluminum #1 to #2, 0.01 to 0.30 mg/L
Type Reagent
compliance-icons compliance-icons
Product Identifier
  • Set Aluminium No. 1/No. 2 100 Pc.
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methenamine