Learn More
Lovibond™ Combi pack aluminum #1 / #2
Click to view available options
Quantity:
100 Tests
Unit Size:
Pack of 100
Description
Tablet reagent test set
Combi pack aluminum #1 / #2 100 tests measuring range 0.01 - 0.30 mg/L ECR method for comparators and photometers.
Specifications
Specifications
| Quantity | 100 Tests |
| Range | Aluminum #1 to #2, 0.01 to 0.30 mg/L |
| Type | Reagent |
Safety and Handling
Product Identifier
- Set Aluminium No. 1/No. 2 100 Pc.
- Warning
- Flammable solid Category 2
- Skin sensitiser Category 1
- H228-Flammable solid.
- H317-May cause an allergic skin reaction.
- EUH210-Safety data sheet available on request.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : methenamine