Learn More
B Braun™ Combifix™ Luer female/Rekord female Adapter
Product Code. 10477902
Change view
Click to view available options
:
Combifix™ Luer female/Rekord female Adapter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10477902 Combifix™ Luer female/Rekord female Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10477902 Supplier B Braun™ Supplier No. 4090306
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Join different and non-standard fitting. B Braun™Combifix™ Luer female/Rekord female Adapter are used for ocnnecting items such as two syringes.
  • Latex-free
  • DEHP free
  • PVC free
  • Transparent
  • Luer female/Rekord female Adapter
Specifications
Color Transparent
Product Type Luer female/Rekord female