Learn More
Xylem Analytics™ WTW™ Type-800 Combined Ion-Selective Electrodes (ISEs)
Click to view available options
Electrode:
Bromide Combined ISE
Calcium Combined ISE
Chloride Combined ISE
Copper Combined ISE
F-, Al, PO¾-, Li+ Combined ISE
Nitric Oxide Combined ISE
Potassium Combined ISE
Silver/Sulfide Combined ISE
Measurement Range:
0.0006 to 6400 mg/L
0.003 to 32,000 mg/L
0.02 mg/L-saturation
0.02 to 40,000 mg/L
0.04 to 39,000 mg/L
0.4 to 62,000 mg/L
0.4 to 79,000 mg/L
2 to 35,000 mg/L
Unit Size:
Each
Description
- Product Type: Type-800 Combined Electrodes
- Use: Measurement of Electrochemical Ion Concentrations
- Connector: DIN or BNC
- Cable Length: 1 m (3.3 ft.)
Specifications
Specifications
| Connector Types | DIN |
| Description | Requirements: ELY/BR/503 reference electrolyte, ionic strength adapter ISA/FK, standard solution ES/Br |
| Length (Metric) Cable | 1 m (3.3 ft.) |
| pH Range | 1 to 12 |
| Model | Br 800 DIN |
| Electrode | Bromide Combined ISE |
| Type | Combination ISE |
| Measurement Range | 0.4 to 79,000 mg/L |