missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Comb 1.5mm

Manufactured to the highest standard. Fisherbrand™ Comb 1.5mm is used with electrophoresis systems.

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11744296

Quantity Price
1 € 57.31 / Each
10 € 49.77 / Each
Estimated Shipment
15-03-2021
Add to basket


Description

Description

  • Used with vertical electrophoresis systems
Specifications

Specifications

Vertical Gel Electrophoresis
48
1.5 mm
Comb
1 Each
200 mm
Videos
SDS
Documents