missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for omniPAGE Mini Vertical Gel Electrophoresis Unit, 0.5mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11887593

Quantity Price
1 € 18.75 / Each
10 € 17.79 / Each
Estimated Shipment: 11-08-2021

Add to basket


Description

Description

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
OmniPAGE Mini Vertical Gel Electrophoresis Unit
16
0.5 mm
Purple
Comb
1 Each
12.5 μL
Videos
SDS
Documents
Certificates