Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
€ 18.85 - € 19.25
Specifications
Color | Black |
---|---|
For Use With (Application) | Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations |
For Use With (Equipment) | OmniPAGE Mini Vertical Gel Electrophoresis Unit |
Type | Comb |
Quantity | 1 Each |
Product Code | Brand | Volume (Metric) Well | No. of Wells | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Well | No. of Wells | Price | Quantity & Availability | ||||
11847583
|
Fisherbrand™
|
500 μL | 1 Prep, 1 Marker |
€ 18.85
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
11867583
|
Fisherbrand™
|
30 μL | 10 |
€ 19.25
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
11847593
|
Fisherbrand™
|
28 μL | 12 |
€ 19.25
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
11897593
|
Fisherbrand™
|
20 μL | 16 |
€ 18.85
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
11847603
|
Fisherbrand™
|
15 μL | 20 |
€ 18.85
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11897603
|
Fisherbrand™
|
70 μL | 5 |
€ 18.85
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
11847613
|
Fisherbrand™
|
40 μL | 8 |
€ 18.85
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
11897613
|
Fisherbrand™
|
35 μL | 9 |
€ 18.85
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
Description
- Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
- Recommended for ultra resolved bands
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Black | |
OmniPAGE Mini Vertical Gel Electrophoresis Unit | |
1 Each |
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations | |
Comb | |
0.75 mm |