Learn More
Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 2mm thick
Click to view available options
No. of Wells:
10
12
16
24
28
Width (Metric) Teeth:
3 mm
5.5 mm
8 mm
9.5 mm
Volume (Metric) Sample:
25 μL
50 μL
70 μL
85 μL
Unit Size:
Each
Description
- Color coded
- Height adjustable
- Offer complete control over loading volume
- Well depth to maximum 112-sample throughput
Specifications
Specifications
| For Use With (Equipment) | HU13 Midi Horizontal Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 24 |
| Thickness (Metric) | 2 mm |
| Volume (Metric) Sample | 25 μL |
| Width (Metric) Teeth | 3 mm |