missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CometAssay Lysis Solution
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Store bottle at room temperature
- CometAssay™ Lysis Solution contains 1% sodium lauryl sarcosinate which is an irritant (precipitates with long term storage at 2-8 °C)
- In case of eye or skin contact, wash thoroughly under running water
- In case of ingestion, rinse mouth with water and seek medical advice
- CometAssay™ Lysis Solution contains high salt and detergent and is suitable for use with either the alkaline or neutral comet assay protocols
- Designed for use with CometAssay™, FLARE™, and CometChip™ Kits
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
| Formulation | Contains 1% sodium lauryl sarcosinate which is an irritant and precipitates with long term storage at 2-8°C. Contains high salt and detergent and is suitable for use with either the alkaline or neutral comet assay protocols. |
| For Use With (Equipment) | CometAssay™, FLARE™, & CometChip™ Kits |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction