Learn More
Tenak™ Comfort Rack for Chest Freezers, 50 mm Box Height, with Boxes
Product Code. 15250814 Shop All TENAK Products
Click to view available options
Holds:
Boxes
Unit Size:
Each
Product Code. 15250814
Brand: Tenak™ TE24504B
Only null left
Add to basket
Add to basket
Comfort Rack suitable for use with all chest freezer models.
  • Racks with spring locks mounded on each shelf to secure the cryoboxes
  • Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
  • Measure the inside height of your freezer when choosing the right rack
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Includes Spring Locks
Holds Boxes
Material Stainless Steel
Positions 6
For Use With (Equipment) Chest Frseezers
Dimensions (L x W x H) 140 x 140 x 355 mm
Height (Metric) 50 mm (Box)