missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD1 Antibody (OTI2E2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72418AF405
This item is not returnable.
View return policy
Description
COMMD1 Monoclonal specifically detects COMMD1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| COMMD1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| COMMD1 | |
| Full length recombinant protein of human COMMD1 (NP_689729) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2E2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 150684.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction