missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD1 Antibody (OTI2E2), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72418PEATT594
This item is not returnable.
View return policy
Description
COMMD1 Monoclonal specifically detects COMMD1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications
| COMMD1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| COMMD1 | |
| Full length recombinant protein of human COMMD1 (NP_689729) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2a |
| Flow Cytometry | |
| OTI2E2 | |
| Flow Cytometry | |
| C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 150684.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction