missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMMD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598616
This item is not returnable.
View return policy
Description
COMMD1 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Proposed scaffold protein that is implicated in diverse physiological processes and whose function may be in part linked to its ability to regulate ubiquitination of specific cellular proteins. Can modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes by displacing CAND1; in vitro promotes CRL E3 activiy and dissocoiates CAND1 from CUL1 and CUL2. Promotes ubiquitination of NF-kappa-B subunit RELA and its subsequent proteasomal degradation. Down-regulates NF-kappa-B activity. Involved in the regulation of membrane expression and ubiquitination of SLC12A2. Modulates Na(+) transport in epithelial cells by regulation of apical cell surface expression of amiloride-sensitive sodium channel (ENaC) subunits and by promoting their ubiquitination presumably involving NEDD4L. Promotes the localization of SCNN1D to recycling endosomes. Promotes CFTR cell surface expression through regulation of its ubiquitination. Down-regulates SOD1 activity by interfering with its homodimerization. Plays a role in copper ion homeostasis. Involved in copper-dependent ATP7A trafficking between the trans- Golgi network and vesicles in the cell periphery; the function is proposed to depend on its association within the CCC complex and cooperation with the WASH complex on early endosomes. Can bind one copper ion per monomer. May function to facilitate biliary copper excretion within hepatocytes. Binds to phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). Involved in the regulation of HIF1A-mediated transcription; competes with ARNT/Hif-1-beta for binding to HIF1A resulting in decreased DNA binding and impaired transcriptional activation by HIF-1.
Specifications
| COMMD1 | |
| Polyclonal | |
| Unconjugated | |
| COMMD1 | |
| AI256843; C2orf5; COMM domain containing 1; COMM domain-containing protein 1; Commd1; copper metabolism (Murr1) domain containing 1; copper metabolism domain containing 1; copper metabolism gene MURR1; Murr1; Protein Murr1; U2/Mu; U2af1-rs1 region 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150684 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N668 | |
| COMMD1 | |
| Recombinant Human COMM domain-containing protein 1 protein (1-190AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction