missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD10 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97236PE
This item is not returnable.
View return policy
Description
COMMD10 Polyclonal antibody specifically detects COMMD10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| COMMD10 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| COMM domain containing 10, COMM domain-containing protein 10, FLJ11285, PTD002 | |
| Produced in rabbits immunized with E. coli-derived Human COMMD10 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51397 | |
| Store at 4C in the dark. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?