missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ COMMD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30505
This item is not returnable.
View return policy
Description
COMMD4 Polyclonal antibody specifically detects COMMD4 in Human samples. It is validated for Western Blot
Specifications
| COMMD4 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54939 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| COMM domain containing 4, COMM domain-containing protein 4, FLJ20452 | |
| between 149 and 199 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction