Learn More
COMMD9 Antibody (002), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90291C
Product Code. 18259906
  • € 617.00 / 0.10mL
Description
COMMD9 Monoclonal specifically detects COMMD9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COMMD9
Monoclonal
DyLight 650
50mM Sodium Borate
Rabbit
Protein A purified
RUO
29099
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
2
Immunohistochemistry, Immunohistochemistry-Paraffin
COMM domain containing 9, COMM domain-containing protein 9, COMM9, FLJ31106, HSPC166
This antibody was obtained from a rabbit immunized with purified, recombinant Human COMMD9 (Q9P000-1; Met1-Lys198).
0.1 mL