missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD9 Antibody (04), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06601F
This item is not returnable.
View return policy
Description
COMMD9 Monoclonal antibody specifically detects COMMD9 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| COMMD9 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 29099 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| 04 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| COMM domain containing 9, COMM domain-containing protein 9, COMM9, FLJ31106, HSPC166 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human COMMD9 (Q9P000-1; Met1-Lys198). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction