missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common beta Chain Antibody (BION-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09024-0.2mg
This item is not returnable.
View return policy
Description
Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Functional Assay, Neutralization
Specifications
| Common beta Chain | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1439 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Functional Assay, Neutralization | |
| BION-1 | |
| Unconjugated | |
| beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
| BALB/c mice were immunized intraperitonally with COS cells transfected DQP (Domain 4 of the common beta-chain) expression constructs. The immunizations were repeated 4 times at 2-week intervals. Four weeks after the final immunization, a mouse was boosted with COS transfectants intravenously. Three days later, splenocytes were harvested and fused with NS-1 myeloma cells. Hybridoma supernatants were screened on CHO DQP cells by flow cytometry, with untransfected CHO cells as a control. All antibodies were from single hybridoma clones as selected by a limiting dilution method. MoAbs were purified from ascites fluid or hybridoma supernatant by a protein A sepharose column. | |
| 0.2 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction