missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common beta Chain Antibody (OTI4H3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70463AF700
This item is not returnable.
View return policy
Description
Common beta Chain Monoclonal specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Common beta Chain | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CSF2RB | |
| Full length recombinant protein of human Common beta Chain (NP_000386) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4H3 | |
| Western Blot, Immunohistochemistry | |
| beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific | |
| 1439.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction