Learn More
Thermo Scientific™ Temperature Control Connectivity Accessories
Product Code. 11608048 Shop All Thermo Scientific Products
Click to view available options
Type:
1.8 m Interface Cable
5 ft. Interface Cable
5 m Interface Cable
Communication Extension Board
Software
For Use With (Equipment):
Cold/Heated Bath Circulators
RS-232 Communications Interface
Recirculating Chiller or Bath
Unit Size:
Each
Product Code. 11608048
Brand: Thermo Scientific™ 422000000004
Only null left
Add to basket
Add to basket
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
  • Choose upper and lower temperature alarm limits and monitor system status with an alarm
  • Record your results on a user selectable graph
  • Select from easy–to–use product icons
  • Create graphs and charts
  • Easily configure ramping functions over time
  • View pop-up alarm windows which display the error condition
  • View a virtual controller screen
  • PC compatible only
TRUSTED_SUSTAINABILITY
Specifications
Communications RS232
For Use With (Equipment) RS-232 Communications Interface
Type Software