Learn More
Thermo Scientific™ Temperature Control Connectivity Accessories
Shop All Thermo Scientific Products
Click to view available options
Type:
1.8 m Interface Cable
5 ft. Interface Cable
5 m Interface Cable
Communication Extension Board
Software
For Use With (Equipment):
Cold/Heated Bath Circulators
RS-232 Communications Interface
Recirculating Chiller or Bath
Unit Size:
Each
Description
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.
- Available for temperature control devices equipped with communications
- Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
- Available in popular configurations that match your device’s connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller or bath
- Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
- Choose upper and lower temperature alarm limits and monitor system status with an alarm
- Record your results on a user selectable graph
- Select from easy–to–use product icons
- Create graphs and charts
- Easily configure ramping functions over time
- View pop-up alarm windows which display the error condition
- View a virtual controller screen
- PC compatible only
Specifications
Specifications
| Communications | RS232, RS485 |
| For Use With (Equipment) | Cold/Heated Bath Circulators |
| Type | 5 ft. Interface Cable |