missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMP/Thrombospondin-5 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99528AF647
This item is not returnable.
View return policy
Description
COMP/Thrombospondin-5 Polyclonal antibody specifically detects COMP/Thrombospondin-5 in Mouse samples. It is validated for Western Blot
Specifications
| COMP/Thrombospondin-5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| cartilage oligomeric matrix protein, cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia1, multiple), cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia1, multiple), EDM1, EPD1, MEDMGC131819, MGC149768, PSACH, pseudoachondroplasia (epiphyseal dysplasia 1, multiple), THBS5, Thrombospondin-5, TSP5 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human COMP/Thrombospondin-5 | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
| 1311 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction