Learn More
Socorex™ Compact Dispenser 501 Model
Specifications
Specifications
| Product Type | Compact Dispenser |
| Volume (Metric) | 0.4 to 5 mL |
| Increments | 0.2 mL |
| Model | 501 |
| Volume (Metric) Reservoir | 1000 mL |
| Warranty | 2 Years |