Learn More
Thermo Scientific™ Heat Insulator (2 Blocks)
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Brand: Thermo Scientific™ 88871108
Additional Details : Weight : 0.20000kg
Description
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 yearsSpecifications
Heat Insulator for 2 block | |
LED | |
107 x 143 x 80 mm (4.2 x 5.6 x 3.2 in.) |
Thermo Scientific™ Compact Dry Bath/Block Heater | |
Heat Insulator |