Learn More
Syngene InGenius 5 Compact Gel Imaging System for DNA/RNA
Product Code. 30306009 Shop All Syngene Products
Change view
Click to view available options
Filter Area:
20 x 24 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Area Unit Size
30306009 20 x 24 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30306009 Supplier Syngene Supplier No. IG5
  • ONLINE EXCLUSIVE PRICE
    € 6025.00
    € 5714.12 / Pack of 1
    Save € 310.88
    5% Off
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A 5M pixel camera gives exquisite pixel resolution and unrivalled sensitivity. InGenius 5 uses a f1.4 manual lens for perfect gel imaging. Internal lighting options includes a UV transilluminator (not included) and overhead white light for easy sample positioning and focusing.
  • Compact gel imaging system for imaging DNA/RNA, stain free, visible protein gels and autorads.
  • The InGenius 5 system is a powerful system with a groundbreaking 5M pixel camera that gives exquisite pixel resolution and unrivalled sensitivity in its class.
  • The system uses a F1.4 manual lens to enable perfect gel imaging.
  • The maximum viewing area is 20 x 24 cm, which is very large for such a small, compact unit.
  • A flexible system with internal lighting options including a UV transilluminator (purchased separately) for working with DNA gels, stain free, and autorads.
  • The overhead LED white lighting is included as standard for easy sample positioning and focusing.
  • The InGenius 5 is a versatile system that has been designed for stain free applications. Its camera enables the sample to be exposed for extended periods.
  • Safely excise your DNA bands from agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
  • An intuitive system that permits images to be saved directly to USB or network for easy transfer.
  • Quickly generate quantitative data with GeneTools analysis software.
  • InGenius 5 comes with unlimited copies of GeneTools.
Specifications
Filter Area 20 x 24 cm
For Use With (Application) Gel Imaging
Product Type Gel Imaging System
Display Type None