missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Antibody (8C3), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 568.00
Specifications
| Antigen | Complement C3 |
|---|---|
| Clone | 8C3 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30028620
|
Novus Biologicals
NBP3-26472-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30030758
|
Novus Biologicals
NBP3-26472-100ul |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement C3 Monoclonal antibody specifically detects Complement C3 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Complement C3 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3 | |
| A synthesized peptide derived from Human Complement C3 [UniProt P01024] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8C3 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 718 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title