missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Antibody (F1-8), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46609AF488
This item is not returnable.
View return policy
Description
Complement C3 Monoclonal specifically detects Complement C3 in Human samples. It is validated for ELISA.
Specifications
| Complement C3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C3 | |
| This Complement C3 Antibody (F1-8) was developed against Complement component 3 (C3) purified from human plasma. | |
| 0.1 mL | |
| Primary | |
| Antibody is specific for human complement component 3 (C3/C3b/C3c). | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA | |
| F1-8 | |
| ELISA | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Mouse | |
| Protein A or G purified | |
| Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| 718 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction