missing translation for 'onlineSavingsMsg'
Learn More

Complement C3 Antibody (S06-2C2), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 421.00 - € 645.00

Specifications

Antigen Complement C3
Clone S06-2C2
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18622334
View Documents
Novus Biologicals
NBP3-19586-100ul
100 μg
€ 645.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633124
View Documents Promotion Details
Novus Biologicals
NBP3-19586-50ul
50 μg
€ 421.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Complement C3 Monoclonal antibody specifically detects Complement C3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Complement C3
0.3 mg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3
A synthetic peptide of human Complement C3 (Uniprot # P01024)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S06-2C2
Western Blot
Unconjugated
Rabbit
Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
718
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.