missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3a Antibody (D17/1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23497AF750
This item is not returnable.
View return policy
Description
Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| Complement C3a | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| C3 | |
| Human C3a | |
| Primary | |
| Recognizes an epitope that is present on human C3, C3a and C3a (desArg). Does not cross-react with C4a or C5a. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| D17/1 | |
| Western Blot, ELISA, Sandwich ELISA | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Mouse | |
| 0.1 mL | |
| 718 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction