missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3d Antibody (053A-1149.3.1.4), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65510JF646
This item is not returnable.
View return policy
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Complement C3d | |
| Monoclonal | |
| Janelia Fluor 646 | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Purified C3d from human plasma | |
| 0.1 mL | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 718 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 053A-1149.3.1.4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction