missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3d Mouse anti-Human, Clone: C3D/2891, Novus Biologicals™
Mouse Monoclonal Antibody
€ 519.00
Specifications
| Antigen | Complement C3d |
|---|---|
| Clone | C3D/2891 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Protein Array |
| Applications | ELISA, Peptide Array |
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for ELISA, Protein Array.Specifications
| Complement C3d | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 718 | |
| IgG2b κ | |
| Protein A or G purified |
| C3D/2891 | |
| ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Recombinant full-length human Complement C3d protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title