missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3d Mouse anti-Human, DyLight 488, Clone: 053A-1149.3.1.4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65510G-0.1ML
This item is not returnable.
View return policy
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Complement C3d | |
| Monoclonal | |
| DyLight 488 | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Purified C3d from human plasma | |
| 0.1 mL | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 718 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 053A-1149.3.1.4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction