Learn More
Complement C3d Mouse anti-Human, PE, Clone: 053A-1149.3.1.4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65510PE-0.1ML
Product Code. 18042974
  • € 887.00 / 0.10mL
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement C3d
Monoclonal
PE
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Purified C3d from human plasma
0.1 mL
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience
718
Store at 4°C in the dark.
IgG1
Flow Cytometry
053A-1149.3.1.4
PBS
Mouse
Protein A purified