missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4a Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05665-100ul
This item is not returnable.
View return policy
Description
Complement C4a Polyclonal antibody specifically detects Complement C4a in Human samples. It is validated for Western Blot
Specifications
| Complement C4a | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2, C4, complement component 4A (Rodgers blood group), RG | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1446 of human Complement C4A (NP_009224.2). LTLTKAPVDLLGVAHNNLMAMAQETGDNLYWGSVTGSQSNAVSPTPAPRNPSDPMPQAPALWIETTAYALLHLLLHEGKAEMADQASAWLTRQGSFQGGFRSTQDTVIALDALSAYWIASHTTEERGLNVTLSSTGRNGFKSHALQLNNRQIRGLEEELQFSLGSKINVKVGGNSKGTLKVLRTYNVLDMKNTTCQDLQIEVTVKGHVEYTMEANEDYEDYEYDELPAKDDPDAPLQPVTPLQLFEG | |
| 100 μg | |
| Immunology, Innate Immunity | |
| 720 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction