missing translation for 'onlineSavingsMsg'
Learn More

Complement C4a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-46760

 View more versions of this product

Product Code. 18662966

  • € 593.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Complement C4a Polyclonal antibody specifically detects Complement C4a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Complement C4a
Polyclonal
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
P0C0L5
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2), 40% Glycerol
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2, C4, complement component 4A (Rodgers blood group), RG
This antibody was developed against Recombinant Protein corresponding to amino acids: PGKPYILTVPGHLDEMQLDIQARYIYGKPVQGVAYVRFGLLDEDGKKTFFRGLESQTKLVNGQSHISLSKAEFQDALEKLNMGITD
0.1 mL
Immunology, Innate Immunity
720
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.