missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Complement C4b/C4c Polyclonal Antibody, FITC
GREENER_CHOICE

Product Code. 11585853
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11585853 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11585853 Supplier Invitrogen™ Supplier No. PA128407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-28407 detects C4c / C4b from human samples. Antigen retrieval for IHC (P): Proteinase K 30 min at 37°C.

This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C4b/C4c
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate FITC
Formulation PBS with 0.09% sodium azide
Gene C4B
Gene Accession No. P0C0L5
Gene Alias Acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; c4 complement; C4A; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4B (Chido blood group); CPAMD2; CPAMD3; DAMC-258G8.6; MGC164979; RG
Gene Symbols C4B
Host Species Rabbit
Immunogen C4c complement isolated from complement activated human serum.
Purification Method IgG fraction
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100293534, 721
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.