missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4d Antibody (SPM545), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34421CL1
This item is not returnable.
View return policy
Description
Complement C4d Monoclonal antibody specifically detects Complement C4d in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/Immunofluorescence
Specifications
| Complement C4d | |
| Monoclonal | |
| CoraFluor 1 | |
| C4, C4A, C4A2, C4A3, C4A4, C4A6, C4S, CO4, complement C4A, CPAMD2, RG | |
| Recombinant human Complement Complement C4d protein | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 720 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| SPM545 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction