Learn More
Novus Biologicals™ Complement C5 Antibody (crovalimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28284-100ug
 View more versions of this product
Product Code. 30046121
  • € 326.00 / 100µg
Description
Complement C5 Monoclonal antibody specifically detects Complement C5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement C5
Monoclonal
Unconjugated
P01031
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
crovalimab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822
Complement C5
100 μg
Immunology, Innate Immunity, Stem Cell Markers
727