Learn More
Complement C7 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00202JF549
Product Code. 18478549
  • ONLINE EXCLUSIVE PRICE
    € 430.00
    € 406.35 / 0.10mL
    Save € 23.65
    5% Off
Description
Complement C7 Polyclonal antibody specifically detects Complement C7 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Produced in rabbits immunized with purified, recombinant Human Complement C7 (Accession#: P10643; Met1-Gln843)
RUO
730
Antigen and protein A Affinity-purified