Learn More
Complement C7 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00202JF646
This item is not returnable.
View return policy
Description
Complement C7 Polyclonal antibody specifically detects Complement C7 in Human samples. It is validated for ELISA
Specifications
| Rabbit | |
| Produced in rabbits immunized with purified, recombinant Human Complement C7 (Accession#: P10643; Met1-Gln843) |