missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement Component C1s Antibody (sutimlimab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28049-100ug
This item is not returnable.
View return policy
Description
Complement Component C1s Monoclonal antibody specifically detects Complement Component C1s in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Complement Component C1s | |
| Monoclonal | |
| Unconjugated | |
| P09871 | |
| Human | |
| Protein A purified | |
| RUO | |
| 716 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| sutimlimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| basic proline-rich peptide IB-1, C1 esterase, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
| C1s | |
| 100 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction