missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C2 Antibody (002), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89300F
This item is not returnable.
View return policy
Description
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for ELISA
Specifications
| Complement Component C2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 717 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 002 | |
| ELISA | |
| C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C2 (Met 1-Leu 752; NP_000054.2). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction