missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C3aR Antibody (3G7), DyLight 350 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52674UV
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, CyTOF
Specifications
| Complement Component C3aR | |
| Monoclonal | |
| DyLight 350 | |
| AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
| The 2e loop of the C3a receptor. | |
| 0.1 mL | |
| GPCR | |
| 719 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, ELISA, Immunofluorescence, CyTOF | |
| 3G7 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction