missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C5/C5a Antibody (06), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06182APC
This item is not returnable.
View return policy
Description
Complement Component C5/C5a Monoclonal antibody specifically detects Complement Component C5/C5a in Human samples. It is validated for ELISA
Specifications
| Complement Component C5/C5a | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 727 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 06 | |
| Sandwich ELISA Capture | |
| C5a;C5b;C5D;Complement C5;CPAMD4;ECLZB | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C5/C5a (Accession#: NP_001726.2; Leu679-Arg751). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction